(R)-(-)-Mandelic acid for synthesis, for resolution of racemates, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid

M-Clarity™ quality level = MQ200

8.06914.0025 8.06914.0100 8.06914.0025 8.06914.0100
EM8.06914.0025CS 10257.36 CAD
EM8.06914.0025 EM8.06914.0100 CA8.06914.0025 CA8.06914.0100
(R)-(-)-Mandelic acid for synthesis, for resolution of racemates, Sigma-Aldrich®
(R)-(-)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00064251
CAS Number: 611-71-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 130 - 132 °C
Molar Mass 152.15 g/mol
Bulk Density 670 kg/m3
Flash Point 190 °C
pH 2.1 (60 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR