n-Octane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06910.0100 8.06910.0250 8.06910.1000 8.06910.0100 8.06910.0250 8.06910.1000
EM8.06910.0100CS 1181.84 CAD
EM8.06910.0100 EM8.06910.0250 EM8.06910.1000 CA8.06910.0100 CA8.06910.0250 CA8.06910.1000
n-Octane for synthesis, Sigma-Aldrich®
n-Octane
Formula: H₃C(CH₂)₆CH₃
MDL Number: MFCD00009556
CAS Number: 111-65-9
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.0007 g/l (20 °C)
Melting Point -57 °C
Molar Mass 114.23 g/mol
Boiling Point 125 - 126 °C (1013 hPa)
Vapor Pressure 14 hPa (20 °C)
Flash Point 13 °C
Explosion Limit 0.8 - 6.5 %(V)
Density 0.70 g/cm3 (20 °C)
Ignition Point 210 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR