1-Nonanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Nonan-1-ol, n-Nonanol, 1-Nonyl alcohol, Nonyl alcohol, 1-Hydroxy nonane, n-Nonyl alcohol, Octyl carbinol
M-Clarity™ quality level = MQ200
8.06866.0100
8.06866.0250
8.06866.0100
8.06866.0250
EM8.06866.0100CS
2279.94
CAD
EM8.06866.0100
EM8.06866.0250
CA8.06866.0100
CA8.06866.0250
1-Nonanol for synthesis, Sigma-Aldrich®
1-Nonanol
Formula:
H₃C(CH₂)₇CH₂OH MW: 144.26 g/mol Boiling Pt: 215 Melting Pt: -7 Flash Pt: 76 |
MDL Number:
MFCD00002990 CAS Number: 143-08-8 UN: 3082 Merck Index: 12,06774 |
Specification Test Results
Solubility | 1 g/l (20 °C) |
Melting Point | -7 °C |
Molar Mass | 144.26 g/mol |
Boiling Point | 214 - 216 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 96 °C |
Refractive Index | 1.4334 (20 °C, 589 nm) |
Explosion Limit | 0.80 - 6.10 %(V) |
Density | 0.83 g/cm3 (20 °C) |
Ignition Point | 237 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...