4-Nitrobenzoyl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06772.0005 8.06772.0250 8.06772.0005 8.06772.0250
EM8.06772.0005CS 38297.62 CAD
EM8.06772.0005 EM8.06772.0250 CA8.06772.0005 CA8.06772.0250
4-Nitrobenzoyl chloride for synthesis, Sigma-Aldrich®
4-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
MDL Number: MFCD00007345
CAS Number: 122-04-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 73 °C
Molar Mass 185.56 g/mol
Boiling Point 155 °C (20 hPa)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR