Nickel aluminium alloy, powder for production of Raney-nickel, for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Nickel-aluminum alloy, Raney®-Nickel alloy, Aluminum-nickel alloy

M-Clarity™ quality level = MQ200

8.06749.0005 8.06749.0250 8.06749.1000 8.06749.0005 8.06749.0250 8.06749.1000
EM8.06749.0005CS 75287 CAD
EM8.06749.0005 EM8.06749.0250 EM8.06749.1000 CA8.06749.0005 CA8.06749.0250 CA8.06749.1000
Nickel aluminium alloy, powder for production of Raney-nickel, for synthesis, Sigma-Aldrich®
Nickel aluminium alloy

for the production of Raney nickel
Melting Point : 1460 °C
Bulk Density: 2850 kg/m3
Density: 4.3 g/cm3

Formula: AlNi
MDL Number: MFCD00167392
CAS Number: 12635-27-7
UN: 1396
ADR: 4.3,III

Order Now

Specification Test Results

Assay of Ni 47.0 - 53.0 %
Assay of Al 47.0 - 53.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR