Sodium hydroxymethanesulfinate dihydrate ≥97.0% stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium hydroxymethanesulphinate dihydrate(Chunks or pellets), Formaldehyde sodium sulfoxylate dihydrate, Sodium hydroxymethanesulfinate dihydrate(Chunks or pellets), Sodium formaldehydesulfoxylate dihydrate, Hydroxymethanesulfinic acid sodium salt dihydrate, Sodium formaldehydesulfoxylate hydrate, Hydroxymethanesulphinic acid sodium salt dihydrate, Formaldehyde sodium sulphoxylate dihydrate, Sodium formaldehydesulphoxylate hydrate, Rongalite dihydrate, Sodium formaldehydesulphoxylate dihydrate

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)
M-Clarity™ quality level = MQ200

8.06455.0100 8.06455.9050 8.06455.1000 8.06455.5000 8.06455.9025 8.06455.1000 8.06455.5000 8.06455.9025
CA8.06455.0100EA 45 CAD
CA8.06455.0100 CA8.06455.9050 EM8.06455.1000 EM8.06455.5000 EM8.06455.9025 CA8.06455.1000 CA8.06455.5000 CA8.06455.9025
Sodium hydroxymethanesulfinate dihydrate ≥97.0% stabilised for synthesis, Sigma-Aldrich®
Sodium hydroxymethanesulphinate dihydrate

This material contains approx. 5% sulfit resulting from the production process

Formula: CH₃NaO₃S·2H₂O
MW: 154.12 g/mol
Melting Pt: 63 °C
Density: 1.8 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150598
CAS Number: 6035-47-8
EINECS: 205-739-4

Order Now

Specification Test Results

Assay (iodometric, calc. as dihydrate) ≥ 97.0 %
Iron (Fe) ≤ 0.003 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR