1,4-Naphthoquinone ≥97% (by HPLC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.06215.0005
8.06215.0250
8.06215.1000
8.06215.9025
8.06215.0005
8.06215.0250
8.06215.1000
8.06215.9025
EM8.06215.0005CS
52498.08
CAD
EM8.06215.0005
EM8.06215.0250
EM8.06215.1000
EM8.06215.9025
CA8.06215.0005
CA8.06215.0250
CA8.06215.1000
CA8.06215.9025
1,4-Naphthoquinone ≥97% (by HPLC) for synthesis, Sigma-Aldrich®
1,4-Naphthoquinone
Formula:
C₁₀H₆O₂ Melting Pt: 120…125 °C (subl.) Density: 1.42 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001676 CAS Number: 130-15-4 UN: 2923 ADR: 8,6.1,III |
Specification Test Results
Solubility | 0.09 g/l (25 °C) |
Melting Point | 120 - 125 °C (sublimed) |
Molar Mass | 158.16 g/mol |
Vapor Pressure | 0.026 hPa (50 °C) |
Flash Point | 141 °C |
Density | 1.42 g/cm3 (20 °C) |
pH | 6.1 (10 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...