1,8-Diaminonaphthalene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.06206.0100
8.06206.0500
8.06206.0100
8.06206.0500
EM8.06206.0100CS
2513.78
CAD
EM8.06206.0100
EM8.06206.0500
CA8.06206.0100
CA8.06206.0500
1,8-Diaminonaphthalene for synthesis, Sigma-Aldrich®
1,8-Diaminonaphthalene
Formula:
C₁₀H₁₀N₂ |
MDL Number:
MFCD00004033 CAS Number: 479-27-6 UN: 3259 ADR: 8,III |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | 61 - 65 °C |
Molar Mass | 158.2 g/mol |
Boiling Point | 205 °C (15.6 hPa) |
Vapor Pressure | 195 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...