2-Methoxyethyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid 2-Methoxyethyl ester, Ethylene glycol monomethyl ether acetate, Methyl glycol acetate, Methyl Cellosolve acetate

M-Clarity™ quality level = MQ200

8.06061.0100 8.06061.2500 8.06061.0100 8.06061.1000 8.06061.2500
EM8.06061.0100CS 839.66 CAD
EM8.06061.0100 EM8.06061.2500 CA8.06061.0100 CA8.06061.1000 CA8.06061.2500
2-Methoxyethyl acetate for synthesis, Sigma-Aldrich®
2-Methoxyethyl acetate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 145
Melting Pt: -65
Flash Pt: 49
MDL Number: MFCD00008719
CAS Number: 110-49-6
UN: 1189
ADR: 3 III
Merck Index: 12,06119

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -65 °C
Molar Mass 118.13 g/mol
Boiling Point 144 - 145 °C (1013 hPa)
Vapor Pressure 4.7 hPa (20 °C)
Flash Point 46 °C
Refractive Index 1.4019 (20 °C, 589 nm)
Explosion Limit 1.7 - 8.8 %(V)
Density 1.01 g/cm3 (20 °C)
Ignition Point 380 °C DIN 51794
Evaporation Number 30


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR