(±)-2-Methyl-1-butanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.06031.0500 8.06031.1000 8.06031.0100 8.06031.0500 8.06031.1000
EM8.06031.0100CS 2401.6 CAD
EM8.06031.0100 EM8.06031.0500 EM8.06031.1000 CA8.06031.0100 CA8.06031.0500 CA8.06031.1000
(±)-2-Methyl-1-butanol for synthesis, Sigma-Aldrich®
(±)-2-Methyl-1-butanol
Formula: C₅H₁₂O
MDL Number: MFCD00004743
CAS Number: 137-32-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Solubility 36 g/l (20 °C)
Melting Point -70 °C
Molar Mass 88.15 g/mol
Boiling Point 127 - 129 °C
Vapor Pressure 4.3 hPa (20 °C)
Flash Point 42.5 °C
Refractive Index 1.4107 (20 °C, 589 nm)
Explosion Limit 1.2 - 10.3 %(V)
Density 0.82 g/cm3 (20 °C)
pH 7 (H2O)
Ignition Point 340 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR