Formaldehyde dimethyl acetal for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06017.1000 8.06017.2500 8.06017.1000 8.06017.2500
EM8.06017.1000CS 1360.38 CAD
EM8.06017.1000 EM8.06017.2500 CA8.06017.1000 CA8.06017.2500
Formaldehyde dimethyl acetal for synthesis, Sigma-Aldrich®
Formaldehyde dimethyl acetal
Formula: CH₂(OCH₃)₂
MDL Number: MFCD00008495
CAS Number: 109-87-5
UN: 1234
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7dc068e2803e769f0f748f5152cbff2cbfe69fc&vid=eae965f4719d855b0e2585391d097f07b53b9067&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -105 °C
Molar Mass 76.11 g/mol
Boiling Point 42.3 °C (1013 hPa)
Vapor Pressure 440 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.3541 (20 °C, 589 nm)
Explosion Limit 1.6 - 17.6 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 235 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR