p-Anisic acid ≥98.0% (by HPLC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.05971.9025 8.05971.0250 8.05971.1000 8.05971.9025 8.05971.0250 8.05971.1000
CA8.05971.9025EA 1754 CAD
CA8.05971.9025 EM8.05971.0250 EM8.05971.1000 EM8.05971.9025 CA8.05971.0250 CA8.05971.1000
p-Anisic acid ≥98.0% (by HPLC) for synthesis, Sigma-Aldrich®
p-Anisic acid
Formula: C₈H₈O₃
Boiling Pt: 277 °C (1013 hPa)
Melting Pt: 182…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002542
CAS Number: 100-09-4

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point 182 - 185 °C
Molar Mass 152.15 g/mol
Bulk Density 380 kg/m3
Boiling Point 277 °C (1013 hPa)
pH 3 - 4 (0.3 g/l, H2O, 20 °C)
Ignition Point 185 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR