3-Chloro-2-methylpropene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.05905.0100
8.05905.0500
8.05905.0100
8.05905.0500
EM8.05905.0100CS
1158.14
CAD
EM8.05905.0100
EM8.05905.0500
CA8.05905.0100
CA8.05905.0500
3-Chloro-2-methylpropene for synthesis, Sigma-Aldrich®
3-Chloro-2-methylpropene
Formula:
C₄H₇Cl MW: 90.55 g/mol Boiling Pt: 72 Melting Pt: -80 |
MDL Number:
MFCD00000953 CAS Number: 563-47-3 UN: 2554 ADR: 3 II Merck Index: 13,02167 |
Specification Test Results
Solubility | 0.5 g/l (20 °C) |
Melting Point | -80 °C |
Molar Mass | 90.55 g/mol |
Boiling Point | 72 - 74 °C |
Vapor Pressure | 140 hPa (20 °C) |
Flash Point | -18 °C |
Refractive Index | 1.4272 (20 °C, 589 nm) |
Explosion Limit | 2.2 - 10.4 %(V) |
Density | 0.93 g/cm3 (20 °C) |
Ignition Point | 540 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...