4-Methylmorpholine ≥98% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: NMM, N-Methylmorpholine

M-Clarity™ quality level = MQ200

8.05894.0005 8.05894.0250 8.05894.1000 8.05894.2500 8.05894.9025 8.05894.0005 8.05894.0250 8.05894.1000 8.05894.2500 8.05894.9025
EM8.05894.0005CS 2645.76 CAD
EM8.05894.0005 EM8.05894.0250 EM8.05894.1000 EM8.05894.2500 EM8.05894.9025 CA8.05894.0005 CA8.05894.0250 CA8.05894.1000 CA8.05894.2500 CA8.05894.9025
4-Methylmorpholine ≥98% (by GC) for synthesis, Sigma-Aldrich®
4-Methylmorpholine
Formula: C₅H₁₁NO
Boiling Pt: 112…114 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.92 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006175
CAS Number: 109-02-4
UN: 2535
ADR: 3.8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -65 °C
Molar Mass 101.15 g/mol
Boiling Point 112 - 114 °C
Vapor Pressure 30 hPa (20 °C)
Flash Point 12 °C DIN 51755 Part 1
Refractive Index 1.4349 (20 °C, 589 nm)
Explosion Limit 2.2 - 11.8 %(V)
Density 0.92 g/cm3 (20 °C)
pH 10.6 (50 g/l, H2O, 20 °C)
Ignition Point 165 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR