1-Methylimidazole (NMI) ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI

M-Clarity™ quality level = MQ200

8.05852.0100 8.05852.1000 8.05852.0100 8.05852.1000
EM8.05852.0100CS 1635.3 CAD
EM8.05852.0100 EM8.05852.1000 CA8.05852.0100 CA8.05852.1000
1-Methylimidazole (NMI) ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
1-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -2 °C
Molar Mass 82.10 g/mol
Boiling Point 198 °C (1013 hPa)
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 92 °C
Explosion Limit 2.7 - 15.7 %(V)
Density 1.035 g/cm3 (20 °C)
pH 9.5 - 10.5 (50 g/l, H2O, 20 °C)
Ignition Point 525 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR