Mesityl oxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Methyl-3-penten-2-one

M-Clarity™ quality level = MQ200

8.05791.0100 8.05791.1000 8.05791.0100 8.05791.1000
EM8.05791.0100CS 850.04 CAD
EM8.05791.0100 EM8.05791.1000 CA8.05791.0100 CA8.05791.1000
Mesityl oxide for synthesis, Sigma-Aldrich®
Mesityl oxide
Formula: C₆H₁₀O
MDL Number: MFCD00008900
CAS Number: 141-79-7
UN: 1229
ADR: 3,III

Order Now

Specification Test Results

Solubility 30 g/l (20 °C)
Melting Point -59 °C
Molar Mass 98.14 g/mol
Boiling Point 130 °C
Vapor Pressure 10.5 hPa (20 °C)
Flash Point 28 °C
Refractive Index 1.4445 (20 °C, 589 nm)
Explosion Limit 1.4 - 10.1 %(V)
Density 0.85 g/cm3 (20 °C)
Ignition Point 344 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR