Potassium-tert-butoxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide

M-Clarity™ quality level = MQ200

8.04918.0100 8.04918.2500 8.04918.3700 8.04918.0100 8.04918.0250 8.04918.2500 8.04918.3700
EM8.04918.0100CS 2619.64 CAD
EM8.04918.0100 EM8.04918.2500 EM8.04918.3700 CA8.04918.0100 CA8.04918.0250 CA8.04918.2500 CA8.04918.3700
Potassium-tert-butoxide for synthesis, Sigma-Aldrich®
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
MDL Number: MFCD00012162
CAS Number: 865-47-4
UN: 2921
ADR: 8 (4.1),I

Order Now

Specification Test Results

Solubility (20 °C) Hydrolysis,(rigorous decomposition)
Melting Point 256 - 258 °C
Molar Mass 112.21 g/mol
Bulk Density 500 kg/m3
Boiling Point 220 °C (1 hPa) (sublimed)
Vapor Pressure 0.07 hPa (180 °C)
Density 0.978 g/cm3 (20 °C)
pH 13 (5 g/l, H2O, 20 °C) Hydrolysis
Ignition Point 360 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR