Isophthaloyl dichloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.04808.0100 8.04808.1000 8.04808.5000 8.04808.0100 8.04808.1000 8.04808.5000
EM8.04808.0100CS 1465.66 CAD
EM8.04808.0100 EM8.04808.1000 EM8.04808.5000 CA8.04808.0100 CA8.04808.1000 CA8.04808.5000
Isophthaloyl dichloride for synthesis, Sigma-Aldrich®
Isophthaloyl dichloride
Formula: C₈H₄Cl₂O₂
MDL Number: MFCD00000678
CAS Number: 99-63-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 42 - 45 °C
Molar Mass 203.03 g/mol
Boiling Point 276 °C
Vapor Pressure 0.04 hPa (25 °C)
Flash Point 180 °C
Explosion Limit 1.5 - 8.9 %(V)
Density 1.39 g/cm3 (17 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR