Isatoic anhydride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.04784.0100
8.04784.0500
8.04784.0100
8.04784.0500
EM8.04784.0100CS
1856.5
CAD
EM8.04784.0100
EM8.04784.0500
CA8.04784.0100
CA8.04784.0500
Isatoic anhydride for synthesis, Sigma-Aldrich®
Isatoic anhydride
Formula:
C₈H₅NO₃ |
MDL Number:
MFCD00006700 CAS Number: 118-48-9 |
Specification Test Results
Solubility | 0.3 g/l (20 °C) |
Melting Point | 230 - 232 °C |
Molar Mass | 163.13 g/mol |
Bulk Density | 600 kg/m3 |
Boiling Point | 350 °C (1013 hPa) |
Flash Point | 308 °C |
Ignition Point |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...