N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

M-Clarity™ quality level = MQ200

8.04518.0025 8.04518.0100 8.04518.0500 8.04518.0025 8.04518.0100 8.04518.0500
EM8.04518.0025CS 5639.02 CAD
EM8.04518.0025 EM8.04518.0100 EM8.04518.0500 CA8.04518.0025 CA8.04518.0100 CA8.04518.0500
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
N-Hydroxysuccinimide
Formula: C₄H₅NO₃
MW: 115.09 g/mol
Boiling Pt: 363
MDL Number: MFCD00005516
CAS Number: 6066-82-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 93 - 100 °C
Molar Mass 115.09 g/mol
pH 5 - 7 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR