HMDS (1,1,1,3,3,3-hexamethyldisilazane) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hexamethyldisilazane

M-Clarity™ quality level = MQ200

8.04324.0050 8.04324.0250 8.04324.1000 8.04324.0050 8.04324.0250 8.04324.1000
EM8.04324.0050CS 4831.64 CAD
EM8.04324.0050 EM8.04324.0250 EM8.04324.1000 CA8.04324.0050 CA8.04324.0250 CA8.04324.1000
HMDS (1,1,1,3,3,3-hexamethyldisilazane) for synthesis, Sigma-Aldrich®
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
Boiling Pt: 126
Melting Pt: -82
Flash Pt: 6
MDL Number: MFCD00008259
CAS Number: 999-97-3
UN: 3286
ADR: 3 (6.1, 8) II
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) decomposes
Melting Point -82 °C
Molar Mass 161.39 g/mol
Boiling Point 126 °C (1013 mbar)
Vapor Pressure 20 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 0.8 - 25.9 %(V)
Density 0.78 g/cm3 (20 °C)
Ignition Point 325 °C
Viscosity Kinematic 0.9 mm2/s (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR