1,6-Hexanediol ≥96.5% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Hexamethylene glycol, 1,6-Dihydroxy hexane

M-Clarity™ quality level = MQ200

8.04308.0100 8.04308.1000 8.04308.9050 8.04308.0100 8.04308.1000 8.04308.9050
EM8.04308.0100CS 1112.32 CAD
EM8.04308.0100 EM8.04308.1000 EM8.04308.9050 CA8.04308.0100 CA8.04308.1000 CA8.04308.9050
1,6-Hexanediol ≥96.5% (by GC) for synthesis, Sigma-Aldrich®
1,6-Hexanediol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 253…260 °C
Melting Pt: 37…42 °C
Density: 1.12 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002985
CAS Number: 629-11-8
Merck Index: 13,04709

Order Now

Specification Test Results

Solubility 5000 g/l (20 °C)
Melting Point 39 - 42 °C
Molar Mass 118.17 g/mol
Bulk Density 530 kg/m3
Boiling Point 253 - 260 °C
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 147 °C
Explosion Limit 6.6 - 16 %(V)
Density 1.12 g/cm3 (20 °C)
pH 7.6 (900 g/l, H2O, 20 °C)
Ignition Point 320 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR