Dibutyltin dichloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.04184.0250 8.04184.1000 8.04184.0250 8.04184.1000
EM8.04184.0250CS 6395.84 CAD
EM8.04184.0250 EM8.04184.1000 CA8.04184.0250 CA8.04184.1000
Dibutyltin dichloride for synthesis, Sigma-Aldrich®
Dibutyltin dichloride
Formula: C₈H₁₈Cl₂Sn
MDL Number: MFCD00000518
CAS Number: 683-18-1
UN: 3146
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 0.32 g/l (20 °C)
Melting Point 37 - 41 °C
Molar Mass 303.85 g/mol
Boiling Point 148 °C (16 hPa)
Vapor Pressure 0.0016 hPa (25 °C)
Flash Point 100 °C
Density 1.37 - 1.40 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR