Bis(triphenylphosphine)palladium(II) chloride Pd 15,2% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Bis(triphenylphosphine)palladium(II) dichloride

M-Clarity™ quality level = MQ200

8.04174.0001 8.04174.0005 8.04174.8500 8.04174.0001 8.04174.0005 8.04174.8500
EM8.04174.0001CS 215753.74 CAD
EM8.04174.0001 EM8.04174.0005 EM8.04174.8500 CA8.04174.0001 CA8.04174.0005 CA8.04174.8500
Bis(triphenylphosphine)palladium(II) chloride Pd 15,2% for synthesis, Sigma-Aldrich®
Bis(triphenylphosphine)palladium (II) chloride
Formula: C₃₆H₃₀Cl₂P₂Pd
MW: 701.91 g/mol
MDL Number: MFCD00009593
CAS Number: 13965-03-2
UN: 1759
ADR: 8 III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 701.91 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR