Ferrocene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03978.0050 8.03978.5000 8.03978.0050 8.03978.5000
EM8.03978.0050CS 1322.46 CAD
EM8.03978.0050 EM8.03978.5000 CA8.03978.0050 CA8.03978.5000
Ferrocene for synthesis, Sigma-Aldrich®
Ferrocene
Formula: C₁₀H₁₀Fe
MW: 186.04 g/mol
Boiling Pt: 249
MDL Number: MFCD00001427
CAS Number: 102-54-5
UN: 1325
ADR: 4.1 II
Merck Index: 13,04070

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 172 - 175 °C
Molar Mass 186.03 g/mol
Bulk Density 500 kg/m3
Boiling Point 249 °C
Vapor Pressure 3.4 hPa (100 °C)
Density 1.49 g/cm3 (20 °C)
Ignition Point 150 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR