N,N-Dimethylethylendiamine ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.03779.0100
8.03779.9022
8.03779.0100
8.03779.9022
EM8.03779.0100CS
2559.6
CAD
EM8.03779.0100
EM8.03779.9022
CA8.03779.0100
CA8.03779.9022
N,N-Dimethylethylendiamine ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
N,N-Dimethylethylendiamine
Formula:
C₄H₁₂N₂ Boiling Pt: 107…109 °C (1013 hPa) Melting Pt: <–70 °C Density: 0.81 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008175 CAS Number: 108-00-9 UN: 2920 ADR: 3.8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Molar Mass | 88.15 g/mol |
Boiling Point | 107 - 109 °C (1013 hPa) |
Vapor Pressure | 29 hPa (20 °C) |
Flash Point | 12.5 °C |
Explosion Limit | 1.3 - 10.8 %(V) |
Density | 0.81 g/cm3 (20 °C) |
pH | 11.4 (10 g/l, H2O, 20 °C) |
Ignition Point | 225 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...