2,6-Xylenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene

M-Clarity™ quality level = MQ200

8.03772.0005 8.03772.0250 8.03772.0005 8.03772.0250
EM8.03772.0005CS 51376.86 CAD
EM8.03772.0005 EM8.03772.0250 CA8.03772.0005 CA8.03772.0250
2,6-Xylenol for synthesis, Sigma-Aldrich®
2,6-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 203
Flash Pt: 73
MDL Number: MFCD00002240
CAS Number: 576-26-1
UN: 2261
ADR: 6.1 II
Merck Index: 12,10215

Order Now

Specification Test Results

Solubility 8 g/l (20 °C)
Melting Point 45 - 48 °C
Molar Mass 122.16 g/mol
Boiling Point 203 °C
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 73 °C
Explosion Limit 1.4 %(V)
Density 1.03 g/cm3 (20 °C)
pH 6 - 7 (8 g/l, H2O, 20 °C)
Ignition Point 555 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR