1,5-Diazobicyclo[4.3.0]-5-nonene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03603.0025 8.03603.0100 8.03603.0025 8.03603.0100
EM8.03603.0025CS 8753.2 CAD
EM8.03603.0025 EM8.03603.0100 CA8.03603.0025 CA8.03603.0100
1,5-Diazobicyclo[4.3.0]-5-nonene for synthesis, Sigma-Aldrich®
1,5-Diazobicyclo[4.3.0]-5-nonene
Formula: C₇H₁₂N₂
MDL Number: MFCD00005554
CAS Number: 3001-72-7
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 124.18 g/mol
Boiling Point 110 °C (20 hPa)
Flash Point 94 °C
Refractive Index 1.519 (20 °C, 589 nm)
Density 1.04 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR