1,3-Diphenyl-2-thiourea for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Thiocarbanilide, N,N'-Diphenylthiourea, N,N’-Diphenylthiourea

M-Clarity™ quality level = MQ200

8.03581.0100 8.03581.0500 8.03581.0100 8.03581.0500
EM8.03581.0100CS 959.06 CAD
EM8.03581.0100 EM8.03581.0500 CA8.03581.0100 CA8.03581.0500
1,3-Diphenyl-2-thiourea for synthesis, Sigma-Aldrich®
1,3-Diphenyl-2-thiourea
Formula: C₁₃H₁₂N₂S
MW: 228.32 g/mol
Boiling Pt: 365
MDL Number: MFCD00004921
CAS Number: 102-08-9
UN: 3077
ADR: 9 III
Merck Index: 13,03366

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 150 - 153 °C
Molar Mass 228.31 g/mol
Bulk Density 400 kg/m3
Density 1.30 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR