2,4-Diamino-6-phenyl-1,3,5-triazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Benzoguanamine, 2,4-Diamino-6-phenyl-s-triazine, 6-Phenyl-1,3,5-triazine-2,4-diyldiamine, 6-Phenyl-1,3,5-triazine-2,4-diamine, 2,6-Diamino-4-phenyl-1,3,5-triazine

M-Clarity™ quality level = MQ200

8.03573.0100 8.03573.1000 8.03573.0100 8.03573.1000
EM8.03573.0100CS 10792.98 CAD
EM8.03573.0100 EM8.03573.1000 CA8.03573.0100 CA8.03573.1000
2,4-Diamino-6-phenyl-1,3,5-triazine for synthesis, Sigma-Aldrich®
2,4-Diamino-6-phenyl-1,3,5-triazine
Formula: C₉H₉N₅
MW: 187.2 g/mol
MDL Number: MFCD00023187
CAS Number: 91-76-9

Order Now

Specification Test Results

Solubility 0.3 g/l (25 °C)
Melting Point 224 - 227 °C
Molar Mass 187.21 g/mol
Bulk Density 680 kg/m3
Vapor Pressure (20 °C)
Flash Point 270 °C
Density 1.425 g/cm3 (15 °C)
pH 6.5 (0.3 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR