4,4'-Isopropylidenediphenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03546.0100 8.03546.1000 8.03546.0100 8.03546.1000
EM8.03546.0100CS 758.97 CAD
EM8.03546.0100 EM8.03546.1000 CA8.03546.0100 CA8.03546.1000
4,4'-Isopropylidenediphenol for synthesis, Sigma-Aldrich®
4,4'-Isopropylidenediphenol
Formula: C₁₅H₁₆O₂
MDL Number: MFCD00002366
CAS Number: 80-05-7

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 155 °C
Molar Mass 228.29 g/mol
Bulk Density 600 kg/m3
Boiling Point 220 °C (5 hPa)
Vapor Pressure 0.86 hPa (190 °C)
Flash Point 227 °C
Density 1.19 g/cm3 (25 °C)
pH 6.4 (H2O)
Ignition Point 510 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR