Dioctyltin oxide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Di-n-octyltin oxide, Dioctyloxostannane
M-Clarity™ quality level = MQ200
8.03544.0025
8.03544.0100
8.03544.0025
8.03544.0100
EM8.03544.0025CS
8353.46
CAD
EM8.03544.0025
EM8.03544.0100
CA8.03544.0025
CA8.03544.0100
Dioctyltin oxide for synthesis, Sigma-Aldrich®
Dioctyltin oxide
Formula:
C₁₆H₃₄OSn MW: 361.16 g/mol Boiling Pt: 230 |
MDL Number:
MFCD00013839 CAS Number: 870-08-6 UN: 3146 ADR: 6.1 III |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Molar Mass | 361.15 g/mol |
Bulk Density | 700 kg/m3 |
Boiling Point | 230 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 70 °C |
Ignition Point |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...