Diallylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03521.0250 8.03521.1000 8.03521.0250 8.03521.1000
EM8.03521.0250CS 1414.1 CAD
EM8.03521.0250 EM8.03521.1000 CA8.03521.0250 CA8.03521.1000
Diallylamine for synthesis, Sigma-Aldrich®
Diallylamine
Formula: C₆H₁₁N
MW: 97.16 g/mol
MDL Number: MFCD00008642
CAS Number: 124-02-7
UN: 2359
ADR: 3 (6.1, 8) II

Order Now

Specification Test Results

Solubility 86 g/l (20 °C)
Melting Point -88 °C
Molar Mass 97.15 g/mol
Boiling Point 111 °C (1013 hPa)
Vapor Pressure 24.2 hPa (20 °C)
Flash Point 11 °C
Refractive Index 1.4405 (20 °C, 589 nm)
Explosion Limit 0.93 - 14.2 %(V)
Density 0.79 g/cm3 (20 °C)
pH 11.5 (9.7 g/l, H2O)
Ignition Point 273 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR