1,3-Diethyl-2-thiourea for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N,N'-Diethylthiourea

M-Clarity™ quality level = MQ200

8.03520.0250 8.03520.5000 8.03520.0250 8.03520.5000
EM8.03520.0250CS 1224.5 CAD
EM8.03520.0250 EM8.03520.5000 CA8.03520.0250 CA8.03520.5000
1,3-Diethyl-2-thiourea for synthesis, Sigma-Aldrich®
1,3-Diethyl-2-thiourea
Formula: C₅H₁₂N₂S
MW: 132.23 g/mol
MDL Number: MFCD00004925
CAS Number: 105-55-5

Order Now

Specification Test Results

Solubility 38 g/l (20 °C)
Melting Point 75 - 78 °C
Molar Mass 132.22 g/mol
Bulk Density 735 kg/m3
Flash Point 186 °C
Density 1.12 g/cm3 (20 °C)
pH 7 (38 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR