1,8-Diazabicyclo[5.4.0]undec-7-ene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, DBU

M-Clarity™ quality level = MQ200

8.03282.0025 8.03282.0100 8.03282.2500 8.03282.9050 8.03282.0100 8.03282.2500 8.03282.9050
CA8.03282.0025EA 61 CAD
CA8.03282.0025 EM8.03282.0100 EM8.03282.2500 EM8.03282.9050 CA8.03282.0100 CA8.03282.2500 CA8.03282.9050
1,8-Diazabicyclo[5.4.0]undec-7-ene for synthesis, Sigma-Aldrich®
1,8-Diazabicyclo[5.4.0]undec-7-ene
Formula: C₉H₁₆N₂
Melting Pt: <–70 °C
Density: 1.02 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006930
CAS Number: 6674-22-2
UN: 2922
ADR: 8,6.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 152.24 g/mol
Boiling Point 261 °C (1013 hPa)
Vapor Pressure 0.02 hPa (25 °C)
Flash Point 116 °C
Explosion Limit 1.1 - 6.5 %(V)
Density 1.02 g/cm3 (20 °C)
pH 12.8 (10 g/l, H2O, 20 °C)
Ignition Point 260 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR