2,2'-Phenyliminodiethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03257.0100 8.03257.1000 8.03257.0100 8.03257.1000
EM8.03257.0100CS 9555.84 CAD
EM8.03257.0100 EM8.03257.1000 CA8.03257.0100 CA8.03257.1000
2,2'-Phenyliminodiethanol for synthesis, Sigma-Aldrich®
2,2'-Phenyliminodiethanol
Formula: (C₆H₅)N(CH₂CH₂OH)₂
MDL Number: MFCD00002845
CAS Number: 120-07-0

Order Now

Specification Test Results

Solubility 3 g/l (20 °C)
Melting Point 56 - 59 °C
Molar Mass 181.23 g/mol
Boiling Point 270 °C
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 200 °C
Density 1.1 g/cm3 (60 °C)
pH 8.2 (3 g/l, H2O, 50 °C)
Ignition Point 380 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR