1-(Dimethylamino)-2-propanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N,N-Dimethyl-1-amino-2-propanol, Dimepranol, N,N-Dimethylisopropanolamine

M-Clarity™ quality level = MQ200

8.03247.0100 8.03247.0500 8.03247.0100 8.03247.0500
EM8.03247.0100CS 933.78 CAD
EM8.03247.0100 EM8.03247.0500 CA8.03247.0100 CA8.03247.0500
1-(Dimethylamino)-2-propanol for synthesis, Sigma-Aldrich®
1-(Dimethylamino)-2-propanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
MDL Number: MFCD00004532
CAS Number: 108-16-7
UN: 2734
ADR: 8 (3),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -85 °C
Molar Mass 103.16 g/mol
Boiling Point 125 - 127 °C (1013 hPa)
Vapor Pressure 17.5 hPa (20 °C)
Flash Point 26 °C
Refractive Index 1.4193 (20 °C, 589 nm)
Explosion Limit 2.7 - 11.1 %(V)
Density 0.85 g/cm3 (20 °C)
pH 11 (100 g/l, H2O, 20 °C)
Ignition Point 225 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR