2-(Dimethylamino)-ethanol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Dimethylaminoethanol, N,N-Dimethylethanolamine, DMAE

M-Clarity™ quality level = MQ200

8.03237.0100 8.03237.1000 8.03237.9025 8.03237.0100 8.03237.1000 8.03237.9025
EM8.03237.0100CS 1109.16 CAD
EM8.03237.0100 EM8.03237.1000 EM8.03237.9025 CA8.03237.0100 CA8.03237.1000 CA8.03237.9025
2-(Dimethylamino)-ethanol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
2-Dimethylaminoethanol
Formula: C₄H₁₁NO
Boiling Pt: 132 - 135 °C (1013 hPa)
Melting Pt: –59 °C
Density: 0.89 g/cm3 (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00002846
CAS Number: 108-01-0
EINECS: 203-542-8
UN: 2051
ADR: 8.3,II
Merck Index: 13,02863

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -59 °C
Molar Mass 89.14 g/mol
Boiling Point 132 - 135 °C (1013 hPa)
Vapor Pressure 6.12 hPa (20 °C)
Flash Point 39 °C DIN 51755 Part 1
Refractive Index 1.4294 (20 °C, 589 nm)
Explosion Limit 1.4 - 12.2 %(V)
Density 0.89 g/cm3 (20 °C)
pH 10.5 - 11 (100 g/l, H2O, 20 °C)
Ignition Point 245 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR