1,4-Dichlorobutane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.03227.0250 8.03227.1000 8.03227.0250 8.03227.1000
EM8.03227.0250CS 1019.1 CAD
EM8.03227.0250 EM8.03227.1000 CA8.03227.0250 CA8.03227.1000
1,4-Dichlorobutane for synthesis, Sigma-Aldrich®
1,4-Dichlorobutane
Formula: ClH₂C(CH₂)₂CH₂Cl
MW: 127.01 g/mol
Boiling Pt: 153.9
Melting Pt: -38
Density: 1.1457 g/cm³ (20 °C)
Flash Pt: 53
Storage Temperature: Ambient
MDL Number: MFCD00001011
CAS Number: 110-56-5
EINECS: 203-778-1
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Solubility 2.4 g/l (20 °C)
Melting Point -38 °C
Molar Mass 127.01 g/mol
Boiling Point 155 °C (1013 hPa)
Vapor Pressure 5 hPa (20 °C)
Flash Point 53 °C
Refractive Index 1.4545 (20 °C, 589 nm)
Explosion Limit 1.8 - 8.9 %(V)
Density 1.14 g/cm3 (20 °C)
pH 7.3 (H2O, 20 °C)
Ignition Point 220 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR