Propyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid propyl ester, n-Propyl acetate

M-Clarity™ quality level = MQ200

8.03183.0100 8.03183.0100 8.03183.1000
EM8.03183.0100CS 1083.88 CAD
EM8.03183.0100 CA8.03183.0100 CA8.03183.1000
Propyl acetate for synthesis, Sigma-Aldrich®
Propyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 102
Melting Pt: -95
Density: 0.886 g/cm³ (20 °C)
Flash Pt: 10
Storage Temperature: Ambient
MDL Number: MFCD00009372
CAS Number: 109-60-4
EINECS: 203-686-1
UN: 1276
ADR: 3 II
Merck Index: 12,08026

Order Now

Specification Test Results

Solubility 21.2 g/l (20 °C)
Melting Point -95 °C
Molar Mass 102.13 g/mol
Boiling Point 101.5 °C (1013 hPa)
Vapor Pressure 33 hPa (20 °C)
Flash Point 14 °C
Explosion Limit 1.7 - 8 %(V)
Density 0.89 g/cm3 (20 °C)
Ignition Point 430 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR