3,4-Dihydro-2H-pyran for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.02971.0100 8.02971.0250 8.02971.0100 8.02971.0250
EM8.02971.0100CS 998.56 CAD
EM8.02971.0100 EM8.02971.0250 CA8.02971.0100 CA8.02971.0250
3,4-Dihydro-2H-pyran for synthesis, Sigma-Aldrich®
3,4-Dihydro-2H-pyran
Formula: C₅H₈O
MDL Number: MFCD00006558
CAS Number: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: 3,II

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Melting Point -70 °C
Molar Mass 84.12 g/mol
Boiling Point 89 °C
Vapor Pressure 120 hPa (20 °C)
Flash Point -9 °C
Refractive Index 1.441 (20 °C, 589 nm)
Explosion Limit 1.1 - 13.8 %(V)
Density 0.93 g/cm3 (20 °C)
pH 7 (5 g/l, H2O, 20 °C)
Ignition Point 240 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR