2-Chloropropane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.02809.0100
8.02809.1000
8.02809.2500
8.02809.0100
8.02809.1000
8.02809.2500
EM8.02809.0100CS
1101.26
CAD
EM8.02809.0100
EM8.02809.1000
EM8.02809.2500
CA8.02809.0100
CA8.02809.1000
CA8.02809.2500
2-Chloropropane for synthesis, Sigma-Aldrich®
2-Chloropropane
Formula:
C₃H₇Cl Boiling Pt: 35.74 °C (1013 hPa) Melting Pt: –117 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: –42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000867 CAS Number: 75-29-6 EINECS: 200-858-8 UN: 2356 ADR: 3,I Merck Index: 13,05231 |
Specification Test Results
Solubility | 3.1 g/l (20 °C) |
Melting Point | -117 °C |
Molar Mass | 78.54 g/mol |
Boiling Point | 36 °C (1013 hPa) |
Vapor Pressure | 580 hPa (20 °C) |
Flash Point | -36 °C |
Refractive Index | 1.3778 (20 °C, 589 nm) |
Explosion Limit | 2.8 - 10.7 %(V) |
Density | 0.86 g/cm3 (20 °C) |
Ignition Point | 590 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...