ε-Caprolactam ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam

M-Clarity™ quality level = MQ200

8.02800.0100 8.02800.0500 8.02800.1000 8.02800.9050 8.02800.0100 8.02800.0500 8.02800.1000 8.02800.9050
EM8.02800.0100CS 4387.49 CAD
EM8.02800.0100 EM8.02800.0500 EM8.02800.1000 EM8.02800.9050 CA8.02800.0100 CA8.02800.0500 CA8.02800.1000 CA8.02800.9050
ε-Caprolactam ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
ε-Caprolactam
Formula: C₆H₁₁NO
Boiling Pt: 270.8 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 1.01 g/cm3 at 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00006936
CAS Number: 105-60-2
EINECS: 203-313-2

Order Now

Specification Test Results

Solubility 4560 g/l (20 °C)
Melting Point 69.2 °C
Molar Mass 113.15 g/mol
Bulk Density 500 - 550 kg/m3
Boiling Point 270.8 °C (1013 hPa)
Vapor Pressure 0.0014 hPa (20 °C)
Flash Point 152 °C DIN 51758
Explosion Limit 1.4 - 8 %(V)
Density 1.01 g/cm3 (80 °C)
pH 7.0 - 8.5 (333 g/l, H2O, 20 °C)
Ignition Point 395 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR