4-Chloroaniline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.02613.0100 8.02613.0100 8.02613.0500
EM8.02613.0100CS 311.26 CAD
EM8.02613.0100 CA8.02613.0100 CA8.02613.0500
4-Chloroaniline for synthesis, Sigma-Aldrich®
4-Chloroaniline
Formula: C₆H₆ClN
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 69…71 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Specification Test Results

Solubility 2.2 g/l (25 °C)
Melting Point 66 - 69 °C
Molar Mass 127.57 g/mol
Bulk Density 790 kg/m3
Boiling Point 232 °C (1013 hPa)
Vapor Pressure 0.07 hPa (20 °C)
Flash Point 132 °C DIN 51758
Density 1.17 g/cm3 (70 °C)
pH 6.9 (1 g/l, H2O, 20 °C)
Ignition Point 685 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR