2-Chloroaniline for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.02611.0100
8.02611.0500
8.02611.0100
8.02611.0500
EM8.02611.0100CS
1439.38
CAD
EM8.02611.0100
EM8.02611.0500
CA8.02611.0100
CA8.02611.0500
2-Chloroaniline for synthesis, Sigma-Aldrich®
2-Chloroaniline
Formula:
C₆H₆ClN MW: 127.57 g/mol Boiling Pt: 208 °C (1013 hPa) Melting Pt: –3 °C Density: 1.213 g/cm³ (20 °C) Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007656 CAS Number: 95-51-2 EINECS: 202-426-4 UN: 2019 ADR: 6.1,II Merck Index: 13,02136 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | -3 °C |
Molar Mass | 127.57 g/mol |
Boiling Point | 207 - 209 °C (1013 hPa) |
Vapor Pressure | 0.5 hPa (20 °C) |
Flash Point | 105 °C |
Refractive Index | 1.5877 (20 °C, 589 nm) |
Explosion Limit | 2.4 - 14.2 %(V) |
Density | 1.21 g/cm3 (20 °C) |
pH | (H2O, 20 °C) suspension,weakly alkaline |
Ignition Point |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...