Cobalt(II) chloride, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Cobaltous chloride, Cobalt dichloride

M-Clarity™ quality level = MQ200

8.02540.0025 8.02540.0100 8.02540.0025 8.02540.0100
EM8.02540.0025CS 5716.44 CAD
EM8.02540.0025 EM8.02540.0100 CA8.02540.0025 CA8.02540.0100
Cobalt(II) chloride, anhydrous for synthesis, Sigma-Aldrich®
Cobalt (II) chloride
Formula: Cl₂Co
MW: 129.84 g/mol
Boiling Pt: 1049
Melting Pt: 735
Density: 1.6795 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 12,02498

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 735 °C
Molar Mass 129.83 g/mol
Boiling Point 1049 °C (1013 hPa)
Density 3.36 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR