(+)-Cinchonine for synthesis, for resolution of racemates, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.02506.0025 8.02506.0100 8.02506.0025 8.02506.0100
EM8.02506.0025CS 4324.46 CAD
EM8.02506.0025 EM8.02506.0100 CA8.02506.0025 CA8.02506.0100
(+)-Cinchonine for synthesis, for resolution of racemates, Sigma-Aldrich®
(+)-Cinchonine
Formula: C₁₉H₂₂N₂O
Melting Pt: 261…264 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064372
CAS Number: 118-10-5
EINECS: 204-234-6
Merck Index: 13,02308

Order Now

Specification Test Results

Solubility 0.25 g/l (20 °C)
Melting Point 258 - 264 °C
Molar Mass 294.40 g/mol
Bulk Density 130 kg/m3
pH 9.0 (0.25 g/l, H2O, 20 °C) (as aqueous solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR