4-Chlorobenzaldehyde for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.02453.0005
8.02453.0250
8.02453.1000
8.02453.0005
8.02453.0250
8.02453.1000
EM8.02453.0005CS
50416.22
CAD
EM8.02453.0005
EM8.02453.0250
EM8.02453.1000
CA8.02453.0005
CA8.02453.0250
CA8.02453.1000
4-Chlorobenzaldehyde for synthesis, Sigma-Aldrich®
4-Chlorobenzaldehyde
Formula:
C₇H₅ClO Boiling Pt: 214 °C (1013 hPa) Melting Pt: 46 °C Density: 1.196 g/cm³ (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003379 CAS Number: 104-88-1 EINECS: 203-247-4 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility | 935 mg/l (20 °C) |
Melting Point | 46 °C |
Molar Mass | 140.57 g/mol |
Boiling Point | 214 °C (1013 hPa) |
Vapor Pressure | 0.36 hPa (25 °C) |
Flash Point | 88 °C |
Density | 1.19 g/cm3 (60 °C) |
Ignition Point | 302 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...