Methyl chloroacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.02413.0100 8.02413.1000 8.02413.0100 8.02413.1000
EM8.02413.0100CS 989.08 CAD
EM8.02413.0100 EM8.02413.1000 CA8.02413.0100 CA8.02413.1000
Methyl chloroacetate for synthesis, Sigma-Aldrich®
Methyl chloroacetate
Formula: C₃H₅ClO₂
Boiling Pt: 131 °C (1013 hPa)
Melting Pt: –33 °C
Density: 1.2378 g/cm³ (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00000931
CAS Number: 96-34-4
EINECS: 202-501-1
UN: 2295
ADR: 6.1 (3),I
Merck Index: 12,06122

Order Now

Specification Test Results

Solubility 10 g/l (25 °C)
Melting Point 110 - 113 °C
Molar Mass 108.09 g/mol
Bulk Density 700 kg/m3
Boiling Point 180 °C (sublimed)
Vapor Pressure 0.12 hPa (20 °C)
Density 1.32 g/cm3 (20 °C)
pH 4 (1 g/l, H2O, 20 °C)
Ignition Point 560 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR