p-Benzoquinone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone

M-Clarity™ quality level = MQ200

8.02410.0100 8.02410.1000 8.02410.9025 8.02410.9050 8.02410.0100 8.02410.1000 8.02410.9025 8.02410.9050
EM8.02410.0100CS 851.62 CAD
EM8.02410.0100 EM8.02410.1000 EM8.02410.9025 EM8.02410.9050 CA8.02410.0100 CA8.02410.1000 CA8.02410.9025 CA8.02410.9050
p-Benzoquinone for synthesis, Sigma-Aldrich®
p-Benzoquinone
Formula: C₆H₄O₂
Boiling Pt: ~ 174 °C (1013 hPa)
Melting Pt: 110…113 °C
Density: 1.32 g/cm3 (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001591
CAS Number: 106-51-4
EINECS: 203-405-2
UN: 2587
ADR: 6.1,II
Merck Index: 12,08259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=759b78f70866f47baad25776f322417b64521955&vid=331add73efa00881381cbace038e21cbca76318f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 10 g/l (25 °C)
Melting Point 110 - 113 °C
Molar Mass 108.09 g/mol
Bulk Density 700 kg/m3
Boiling Point 180 °C (sublimed)
Vapor Pressure 0.12 hPa (20 °C)
Density 1.32 g/cm3 (20 °C)
pH 4 (1 g/l, H2O, 20 °C)
Ignition Point 560 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR